| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500021717.mm.1 |
NM_001042620.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021717.mm.1 |
NM_007839.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021717.mm.1 |
NR_104311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021717.mm.1 |
TC0500002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021718.mm.1 |
NM_001042620.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021718.mm.1 |
NM_007839.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021718.mm.1 |
NR_104311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021718.mm.1 |
TC0500002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021720.mm.1 |
NM_001042620.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021720.mm.1 |
NM_007839.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021720.mm.1 |
NR_104311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021720.mm.1 |
TC0500002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021721.mm.1 |
NM_001042620.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021721.mm.1 |
NM_007839.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021721.mm.1 |
NR_104311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021721.mm.1 |
TC0500002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021722.mm.1 |
NM_001042620.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021722.mm.1 |
NM_007839.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021722.mm.1 |
NR_104311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021722.mm.1 |
TC0500002406.mm.1 |
MTA TC |