| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900025386.mm.1 |
NM_008138.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025386.mm.1 |
NM_008138.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025386.mm.1 |
TC0900003038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025384.mm.1 |
NM_008138.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025384.mm.1 |
NM_008138.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025384.mm.1 |
TC0900003038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025387.mm.1 |
NM_008138.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025387.mm.1 |
NM_008138.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025387.mm.1 |
TC0900003038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025385.mm.1 |
NM_008138.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025385.mm.1 |
NM_008138.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025385.mm.1 |
TC0900003038.mm.1 |
MTA TC |