Assay Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163465.1 | NP_001156937.1 | 3 - 4 | 431 | 98 | |
| NM_008164.2 | NP_032190.2 | 3 - 4 | 431 | 98 | ||
| GenBank mRNA | AK143132.1 | - | 3 - 4 | 412 | 98 | |
| AK220431.1 | - | 3 - 4 | 348 | 98 | ||
| BC004843.1 | - | 3 - 4 | 401 | 98 | ||
| BC052010.1 | - | 3 - 4 | 390 | 5717786 | 98 | |
| U43194.1 | - | 3 - 4 | 325 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhpn1
Genomic Map