Assay Details
| Gene Symbol: Icam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008319.2 | NP_032345.2 | 1 - 2 | 180 | 122 | |
| GenBank mRNA | BC113140.1 | - | 1 - 2 | 128 | 40048010 | 122 |
| BY255874.1 | - | 1 - 2 | 168 | 122 | ||
| U06483.1 | - | 1 - 2 | 128 | 122 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC113140.1,NM_008319.2,U06483.1 transcripts.
Set Membership: |
|

Icam5
Genomic Map