Assay Details
| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008378.2 | NP_032404.1 | 4 - 5 | 368 | 119 | |
| GenBank mRNA | AK138136.1 | - | 4 - 5 | 339 | 119 | |
| BC020524.1 | - | 4 - 5 | 344 | 4223620 | 119 | |
| D87973.1 | - | 4 - 5 | 368 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impact
Genomic Map