Assay Details
| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008389.3 | NP_032415.2 | 7 - 8 | 1457 | 70 | |
| GenBank mRNA | AF285178.1 | - | 7 - 8 | 1347 | 70 | |
| AK131718.1 | - | 7 - 8 | 1458 | 70 | ||
| X58523.1 | - | 385 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ipp
Genomic Map