Assay Details
| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008389.3 | NP_032415.2 | 8 - 9 | 1687 | 92 | |
| GenBank mRNA | AF285178.1 | - | 8 - 9 | 1577 | 92 | |
| AK131718.1 | - | 8 - 9 | 1688 | 92 | ||
| X58523.1 | - | 615 | 92 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229578 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | X58523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | X58523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840404 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345787 | AK080723.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345787 | AK220358.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210172 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37432 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141837 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132976 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | X58523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | X58523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05369020 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00966237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05369040 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132976 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05369034 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141837 | - | - |
| Whole Mouse Genome G4122A | A_51_P196455 | AK136451.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196455 | AK042290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | X58523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | X58523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | AF285178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | AK131718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340816 | NM_008389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214460 | AF285178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214460 | AK131718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214460 | NM_008389.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132093 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141837 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05369020 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132976 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05369040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23229580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23229703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04082080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04002782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04042074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03946842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03703533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04002766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03223020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03324179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04002777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03946845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03709194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04082081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03367293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04002781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23229596 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507289 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421771_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507286 | - | - |
| Mouse Genome 430 2.0 Array | 1421771_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421771_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16351_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16351_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400009294.mm.1 | NM_008389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009294.mm.1 | NM_008389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009294.mm.1 | TC0400001253.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400009293.mm.1 | NM_008389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009293.mm.1 | NM_008389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009293.mm.1 | TC0400001253.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421771_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421771_a_at | - | - |
| Murine Genome U74v2 Set | 94699_at | - | - |
| Murine Genome U74v2 Set | 94699_PM_at | - | - |
| Murine Genome U74v2 Set | 171523_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171523_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421771_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421771_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421771_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421771_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 551833 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6020288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237651 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237651 | - | - |
| mouse-6 v1.1 (Illumina) | scl25073.4.1_109-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25073.4.1_109-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237651 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237651 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25073.4.1_109-S | - | - |

Ipp
Genomic Map