Assay Details
| Gene Symbol: Irf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159417.1 | NP_001152889.1 | 7 - 8 | 975 | 69 | |
| NM_001159418.1 | NP_001152890.1 | 6 - 7 | 773 | 69 | ||
| NM_008394.3 | NP_032420.1 | 6 - 7 | 889 | 69 | ||
| GenBank mRNA | AK145911.1 | - | 5 - 6 | 641 | 69 | |
| AK149742.1 | - | 5 - 6 | 640 | 69 | ||
| BC005435.1 | - | 7 - 8 | 750 | 3601255 | 69 | |
| BC012968.1 | - | 6 - 7 | 784 | 3257714 | 69 | |
| U51992.1 | - | 6 - 7 | 693 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf9
Genomic Map