Assay Details
| Gene Symbol: Irf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159417.1 | NP_001152889.1 | 9 - 10 | 1432 | 68 | |
| NM_001159418.1 | NP_001152890.1 | 8 - 9 | 1230 | 68 | ||
| NM_008394.3 | NP_032420.1 | 8 - 9 | 1346 | 68 | ||
| GenBank mRNA | AK145911.1 | - | 7 - 8 | 1098 | 68 | |
| AK149742.1 | - | 7 - 8 | 1098 | 68 | ||
| BC005435.1 | - | 9 - 10 | 1207 | 3601255 | 68 | |
| BC012968.1 | - | 8 - 9 | 1241 | 3257714 | 68 | |
| U51992.1 | - | 8 - 9 | 1150 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114938 | BC012968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114938 | BC005435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114938 | AK145911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114938 | NM_001159418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114938 | NM_008394.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146488 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136413 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114938 | BC012968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114938 | BC005435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114938 | AK145911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114938 | NM_001159418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114938 | NM_008394.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176013 | BC012968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176013 | CK788642.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176013 | U51992.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176013 | AK149742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176013 | BC005435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127367 | BC012968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127367 | U51992.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127367 | AK149742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127367 | BC005435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127367 | AK043697.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29618790 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136413 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08118961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08118971 | - | - |
| Whole Mouse Genome G4122A | A_52_P176013 | BC012968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176013 | CK788642.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176013 | U51992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176013 | AK149742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176013 | BC005435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127367 | BC012968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127367 | U51992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127367 | AK149742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127367 | BC005435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127367 | AK043697.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130883 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136413 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11457289 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11457299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11457297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11457298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29618817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11333706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11338869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11333705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11691490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11457289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11691484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11457283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29618824 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415321 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421322_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415319 | - | - |
| Mouse Genome 430 2.0 Array | 1421322_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421322_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16391_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16391_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_001159417.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_001159417.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_001159418.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_001159418.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_008394.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | NM_008394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006365.mm.1 | TC1400002834.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421322_a_at | - | - |
| Mouse Expression Set 430 | 1421322_PM_a_at | - | - |
| Murine Genome U74v2 Set | 103634_PM_at | - | - |
| Murine Genome U74v2 Set | 103634_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421322_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421322_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421322_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421322_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3190131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3190131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7100142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007100142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2636339 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233461 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2636339 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233461 | - | - |
| mouse-6 v1.1 (Illumina) | scl016391.8_62-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000304.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7100142 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190131 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000304.1_6-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016391.8_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100142 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190131 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2636339 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233461 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2636339 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233461 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl016391.8_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000304.1_6-S | - | - |

Irf9
Genomic Map