Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008437.1 | NP_032463.1 | 4 - 5 | 621 | 70 | |
| GenBank mRNA | AB038144.1 | - | 4 - 5 | 634 | 70 | |
| AK002520.1 | - | 4 - 5 | 468 | 70 | ||
| AK007861.1 | - | 4 - 5 | 467 | 70 | ||
| AK152663.1 | - | 4 - 5 | 459 | 70 | ||
| BC014813.1 | - | 4 - 5 | 454 | 4221844 | 70 | |
| D88899.1 | - | 4 - 5 | 621 | 70 | ||
More Information
Set Membership: |
|

Napsa
Genomic Map