Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008437.1 | NP_032463.1 | 5 - 6 | 825 | 132 | |
| GenBank mRNA | AB038144.1 | - | 5 - 6 | 838 | 132 | |
| AK002520.1 | - | 5 - 6 | 672 | 132 | ||
| AK152663.1 | - | 5 - 6 | 663 | 132 | ||
| AK170013.1 | - | 5 - 6 | 669 | 132 | ||
| BC014813.1 | - | 5 - 6 | 658 | 4221844 | 132 | |
| D88899.1 | - | 5 - 6 | 825 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98264 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131763 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106790 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | D88899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | D88899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25088203 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131763 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106790 | - | - |
| Whole Mouse Genome G4122A | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | AK170013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | BC014813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | AK152663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | D88899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146149 | NM_008437.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132495 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130819 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106790 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131763 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05918595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06643276 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06587153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06587150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25088330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06660493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05958204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06660494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25088246 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552704 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430744_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423590_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552697 | - | - |
| Mouse Genome 430 2.0 Array | 1423590_at | - | - |
| Mouse Genome 430 2.0 Array | 1423590_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430744_at | - | - |
| Mouse Genome 430 2.0 Array | 1430744_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16541_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16541_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006727.mm.1 | NM_008437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006727.mm.1 | NM_008437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006727.mm.1 | TC0700000810.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423590_at | - | - |
| Mouse Expression Set 430 | 1423590_PM_at | - | - |
| Mouse Expression Set 430 | 1430744_PM_at | - | - |
| Mouse Expression Set 430 | 1430744_at | - | - |
| Murine Genome U74v2 Set | 101972_at | - | - |
| Murine Genome U74v2 Set | 101972_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423590_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423590_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423590_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423590_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430744_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430744_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5910364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230302 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657867 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657867 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230302 | - | - |
| mouse-6 v1.1 (Illumina) | scl016541.8_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000223.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910364 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016541.8_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000223.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910364 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810711 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230302 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2781030 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2657867 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657867 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230302 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2781030 | - | - |

Napsa
Genomic Map