Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008437.1 | NP_032463.1 | 7 - 8 | 1088 | 73 | |
| GenBank mRNA | AB038144.1 | - | 7 - 8 | 1101 | 73 | |
| AK002520.1 | - | 7 - 8 | 935 | 73 | ||
| AK152663.1 | - | 7 - 8 | 926 | 73 | ||
| AK170013.1 | - | 7 - 8 | 932 | 73 | ||
| BC014813.1 | - | 7 - 8 | 921 | 4221844 | 73 | |
| D88899.1 | - | 7 - 8 | 1088 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Napsa
Genomic Map