Assay Details
| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008535.2 | NP_032561.2 | 3 - 4 | 977 | 58 | |
| GenBank mRNA | AK076114.1 | - | 3 - 4 | 631 | 58 | |
| BC005736.1 | - | 3 - 4 | 597 | 3598911 | 58 | |
| CT010167.1 | - | 2 - 3 | 422 | 58 | ||
| X57687.1 | - | 3 - 4 | 887 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel maturation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
B cell differentiation
positive regulation of transcription, DNA-templated
definitive hemopoiesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
blood vessel maturation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
B cell differentiation
positive regulation of transcription, DNA-templated
definitive hemopoiesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021285 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152937 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114662 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | X57687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | X57687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26021249 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114662 | - | - |
| Whole Mouse Genome G4122A | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | X57687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | AK076114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | BC005736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | CO044699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143190 | NM_008535.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133400 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07422401 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07422410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07422412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07422405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07422394 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021285 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26021245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26021272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07422394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07422391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06946432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07055400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07422408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07055413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26021268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573423 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419120_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573419 | - | - |
| Mouse Genome 430 2.0 Array | 1419120_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419120_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17095_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17095_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800007259.mm.1 | NM_008535.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007259.mm.1 | NM_008535.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007259.mm.1 | TC0800000959.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419120_at | - | - |
| Mouse Expression Set 430 | 1419120_PM_at | - | - |
| Murine Genome U74v2 Set | 100467_at | - | - |
| Murine Genome U74v2 Set | 100468_PM_g_at | - | - |
| Murine Genome U74v2 Set | 100468_g_at | - | - |
| Murine Genome U74v2 Set | 100467_PM_at | - | - |
| Murine Genome U74v2 Set | 166994_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166994_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419120_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419120_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419120_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419120_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 490946 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2900242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612255 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2612255 | - | - |
| mouse-6 v1.1 (Illumina) | scl33579.4.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900242 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33579.4.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900242 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2612255 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612255 | - | - |

Lyl1
Genomic Map