Assay Details
| Gene Symbol: Lypla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008866.2 | NP_032892.1 | 4 - 5 | 228 | 80 | |
| GenBank mRNA | AK002674.1 | - | 4 - 5 | 232 | 80 | |
| AK050549.1 | - | 4 - 5 | 205 | 80 | ||
| AK146874.1 | - | 4 - 5 | 243 | 80 | ||
| AK167231.1 | - | 250 | 80 | |||
| BC013536.1 | - | 4 - 5 | 228 | 4240573 | 80 | |
| BC052848.1 | - | 4 - 5 | 269 | 30055025 | 80 | |
| CT010201.1 | - | 4 - 5 | 207 | 80 | ||
| U89352.1 | - | 4 - 5 | 207 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lypla1
Genomic Map