| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600019397.mm.1 |
NM_001079822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019397.mm.1 |
NM_009332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019397.mm.1 |
TC0600002496.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019396.mm.1 |
NM_001079822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019396.mm.1 |
NM_009332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019396.mm.1 |
TC0600002496.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019398.mm.1 |
NM_001079822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019398.mm.1 |
NM_009332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019398.mm.1 |
TC0600002496.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019400.mm.1 |
NM_001079822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019400.mm.1 |
NM_009332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019400.mm.1 |
TC0600002496.mm.1 |
MTA TC |