Assay Details
| Gene Symbol: Tcf7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009332.3 | NP_033358.2 | 4 - 5 | 612 | 59 | |
| GenBank mRNA | AJ223069.1 | - | 4 - 5 | 520 | 59 | |
| AJ223233.1 | - | 4 - 5 | 435 | 59 | ||
| AK052721.1 | - | 4 - 5 | 822 | 59 | ||
| BC128306.1 | - | 4 - 5 | 519 | 40109613 | 59 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Citations (1)
Tcf3 is an integral component of the core regulatory circuitry of embryonic stem cells.
- Authors
- Cole MF, Johnstone SE, Newman JJ, Kagey MH, Young RA
- Journal
- Genes Dev - Page No 746 (Sat Mar 15 00:00:00 UTC 2008)
- Volume (Year)
- 22 (2008)
- Product Usage
- -

Tcf7l1
Genomic Map