Assay Details
| Gene Symbol: Tead1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166584.1 | NP_001160056.1 | 12 - 13 | 1552 | 89 | |
| NM_001166585.1 | NP_001160057.1 | 9 - 10 | 1070 | 89 | ||
| NM_009346.3 | NP_033372.1 | 11 - 12 | 1489 | 89 | ||
| GenBank mRNA | AK140307.1 | - | 12 - 13 | 1553 | 89 | |
| AK148375.1 | - | 11 - 12 | 1455 | 89 | ||
| BC052021.1 | - | 9 - 10 | 1011 | 89 | ||
| BC060138.1 | - | 9 - 10 | 1011 | 6410573 | 89 | |
| L06865.1 | - | 9 - 10 | 1016 | 89 | ||
| L13853.1 | - | 11 - 12 | 1216 | 89 | ||
| S74227.1 | - | 10 - 11 | 1477 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead1
Genomic Map