Assay Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009420.2 | NP_033446.1 | 3 - 4 | 255 | 113 | |
| GenBank mRNA | AK018856.1 | - | 3 - 4 | 186 | 113 | |
| BC049615.1 | - | 3 - 4 | 207 | 6742033 | 113 | |
| M25533.1 | - | 3 - 4 | 207 | 113 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK018856.1,BC049615.1,M25533.1,NM_009420.2 transcripts.
Set Membership: |
|

Crisp2
Genomic Map