Assay Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204071.1 | NP_001191000.1 | 9 - 10 | 846 | 114 | |
| NM_009420.2 | NP_033446.1 | 9 - 10 | 864 | 114 | ||
| GenBank mRNA | BC049615.1 | - | 9 - 10 | 816 | 6742033 | 114 |
| M25533.1 | - | 9 - 10 | 816 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | M25533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | M25533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226712 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106155 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110980 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101526 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | M25533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | M25533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31177434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110980 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101526 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08776862 | - | - |
| Whole Mouse Genome G4122A | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | M25533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | BC049615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | CJ046590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | M25533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | NM_001204071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P43282 | NM_009420.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115816 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101526 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08776854 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13114973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13114979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13207545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13114976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13207559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13207554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31177467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13559257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13207546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31177425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13279871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450941 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416717_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450938 | - | - |
| Mouse Genome 430 2.0 Array | 1416717_at | - | - |
| Mouse Genome 430 2.0 Array | 1416717_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22024_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22024_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa12460_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019725.mm.1 | NM_001204071.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019725.mm.1 | NM_001204071.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019725.mm.1 | NM_009420.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019725.mm.1 | NM_009420.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019725.mm.1 | TC1700002106.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416717_at | - | - |
| Mouse Expression Set 430 | 1416717_PM_at | - | - |
| Murine Genome U74v2 Set | 92825_at | - | - |
| Murine Genome U74v2 Set | 92825_PM_at | - | - |
| Murine Genome U74v2 Set | 162454_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162454_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416717_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416717_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416717_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416717_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 372486 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5890670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2715114 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2715114 | - | - |
| mouse-6 v1.1 (Illumina) | scl49911.11.1_99-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49911.11.1_99-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890670 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715114 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2715114 | - | - |

Crisp2
Genomic Map