Assay Details
| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293561.1 | NP_001280490.1 | 6 - 7 | 1090 | 72 | |
| NM_009472.4 | NP_033498.1 | 6 - 7 | 1090 | 72 | ||
| GenBank mRNA | AK031655.1 | - | 6 - 7 | 1057 | 72 | |
| AK142411.1 | - | 6 - 7 | 1056 | 72 | ||
| AK142974.1 | - | 6 - 7 | 984 | 72 | ||
| AK164239.1 | - | 5 - 6 | 834 | 72 | ||
| BC115772.1 | - | 6 - 7 | 1080 | 40085998 | 72 | |
| U72634.1 | - | 6 - 7 | 1090 | 72 | ||
More Information
Set Membership: |
|

Unc5c
Genomic Map