Assay Details
| Gene Symbol: Yap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171147.1 | NP_001164618.1 | 1 - 2 | 480 | 62 | |
| NM_009534.3 | NP_033560.1 | 1 - 2 | 480 | 62 | ||
| GenBank mRNA | AK157234.1 | - | 1 - 2 | 480 | 62 | |
| BC014733.1 | - | 1 - 2 | 477 | 4239820 | 62 | |
| BC039125.1 | - | 1 - 2 | 335 | 2649070 | 62 | |
| BC094313.1 | - | 1 - 2 | 468 | 5697882 | 62 | |
| X80508.1 | - | 1 - 2 | 472 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yap1
Genomic Map