| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00004217.mm.1 |
NM_001109043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004217.mm.1 |
NM_001109043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004217.mm.1 |
NM_009549.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004217.mm.1 |
NM_009549.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004217.mm.1 |
TC0X00000670.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00004215.mm.1 |
NM_001109043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004215.mm.1 |
NM_001109043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004215.mm.1 |
NM_009549.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004215.mm.1 |
NM_009549.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004215.mm.1 |
TC0X00000670.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00004218.mm.1 |
NM_001109043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004218.mm.1 |
NM_001109043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004218.mm.1 |
NM_009549.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004218.mm.1 |
NM_009549.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00004218.mm.1 |
TC0X00000670.mm.1 |
MTA TC |