Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 8 - 9 | 739 | 65 | |
| NM_009549.3 | NP_033575.3 | 8 - 9 | 739 | 65 | ||
| GenBank mRNA | AK048188.1 | - | 8 - 9 | 720 | 65 | |
| BC039772.1 | - | 5 - 6 | 324 | 4191053 | 65 | |
| EF494744.1 | - | 8 - 9 | 719 | 65 | ||
| U46687.1 | - | 7 - 8 | 565 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map