Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 10 - 11 | 859 | 86 | |
| NM_009549.3 | NP_033575.3 | 10 - 11 | 862 | 86 | ||
| GenBank mRNA | AK048188.1 | - | 10 - 11 | 840 | 86 | |
| BC039772.1 | - | 7 - 8 | 447 | 4191053 | 86 | |
| EF494744.1 | - | 10 - 11 | 842 | 86 | ||
| U46687.1 | - | 9 - 10 | 688 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map