Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 13 - 14 | 1225 | 63 | |
| NM_009549.3 | NP_033575.3 | 13 - 14 | 1228 | 63 | ||
| GenBank mRNA | AK014497.1 | - | 2 - 3 | 276 | 63 | |
| AK048188.1 | - | 13 - 14 | 1206 | 63 | ||
| BC039772.1 | - | 10 - 11 | 813 | 4191053 | 63 | |
| EF494744.1 | - | 13 - 14 | 1208 | 63 | ||
| U46687.1 | - | 12 - 13 | 1054 | 63 | ||
More Information
Set Membership: |
|

Zfp185
Genomic Map