Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 16 - 17 | 1485 | 103 | |
| NM_009549.3 | NP_033575.3 | 16 - 17 | 1488 | 103 | ||
| GenBank mRNA | AK014497.1 | - | 5 - 6 | 536 | 103 | |
| AK048188.1 | - | 16 - 17 | 1466 | 103 | ||
| BC039772.1 | - | 13 - 14 | 1073 | 4191053 | 103 | |
| EF494744.1 | - | 16 - 17 | 1468 | 103 | ||
| U46687.1 | - | 15 - 16 | 1314 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map