Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009581.2 | NP_033607.2 | 4 - 5 | 456 | 76 | |
| GenBank mRNA | BB615894.1 | - | 4 - 5 | 456 | 76 | |
| BC119121.1 | - | 4 - 5 | 446 | 8733870 | 76 | |
| BC119123.1 | - | 4 - 5 | 446 | 8733872 | 76 | |
| U17108.1 | - | 4 - 5 | 454 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zp3r
Genomic Map