Assay Details
| Gene Symbol: Ifnar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010509.2 | NP_034639.2 | 7 - 8 | 1040 | 97 | |
| GenBank mRNA | AF013274.1 | - | 7 - 8 | 818 | 97 | |
| AK139169.1 | - | 7 - 8 | 1020 | 97 | ||
| AK150386.1 | - | 7 - 8 | 974 | 97 | ||
| AK150597.1 | - | 7 - 8 | 953 | 97 | ||
| AK151474.1 | - | 7 - 8 | 968 | 97 | ||
| AK151493.1 | - | 7 - 8 | 953 | 97 | ||
| AK151777.1 | - | 7 - 8 | 968 | 97 | ||
| AK152878.1 | - | 7 - 8 | 974 | 97 | ||
| AK153317.1 | - | 7 - 8 | 974 | 97 | ||
| BC071225.1 | - | 6 - 7 | 738 | 4187603 | 97 | |
| Y09865.1 | - | 7 - 8 | 930 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar2
Genomic Map