Assay Details
| Gene Symbol: Krt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010662.1 | NP_034792.1 | 1 - 2 | 468 | 91 | |
| GenBank mRNA | AK004635.1 | - | 1 - 2 | 540 | 91 | |
| AK010156.1 | - | 1 - 2 | 540 | 91 | ||
| AK132640.1 | - | 1 - 2 | 540 | 91 | ||
| BC060286.1 | - | 1 - 2 | 483 | 30292772 | 91 | |
| X03492.1 | - | 1 - 2 | 93 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt13
Genomic Map