Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017959.2 | NP_001017959.1 | 2 - 3 | 323 | 87 | |
| NM_001290485.1 | NP_001277414.1 | 2 - 3 | 323 | 87 | ||
| NM_010685.4 | NP_034815.2 | 2 - 3 | 323 | 87 | ||
| GenBank mRNA | AK012149.1 | - | 2 - 3 | 282 | 87 | |
| AK032974.1 | - | 2 - 3 | 285 | 87 | ||
| AK048174.1 | - | 2 - 3 | 265 | 87 | ||
| AK051989.1 | - | 1 - 2 | 1121 | 87 | ||
| AK078193.1 | - | 2 - 3 | 317 | 87 | ||
| AK088804.1 | - | 3 - 4 | 300 | 87 | ||
| AK136550.1 | - | 2 - 3 | 284 | 87 | ||
| AK146731.1 | - | 2 - 3 | 304 | 87 | ||
| AK150766.1 | - | 2 - 3 | 292 | 87 | ||
| AK159272.1 | - | 2 - 3 | 278 | 87 | ||
| AK159731.1 | - | 2 - 3 | 278 | 87 | ||
| AK163933.1 | - | 2 - 3 | 324 | 87 | ||
| BC138718.1 | - | 2 - 3 | 210 | 8861740 | 87 | |
| BC138723.1 | - | 2 - 3 | 210 | 8861745 | 87 | |
| BY283193.1 | - | 2 - 3 | 318 | 87 | ||
| J05287.1 | - | 2 - 3 | 250 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map