Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159301.1 | NP_001152773.1 | 7 - 8 | 676 | 79 | |
| NM_010708.2 | NP_034838.2 | 8 - 9 | 769 | 79 | ||
| GenBank mRNA | AK146044.1 | - | 7 - 8 | 632 | 79 | |
| AK214388.1 | - | 2 - 3 | 67 | 79 | ||
| BC003754.1 | - | 7 - 8 | 603 | 3601419 | 79 | |
| U55060.1 | - | 7 - 8 | 602 | 79 | ||
| U55061.1 | - | 8 - 9 | 676 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals9
Genomic Map