| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_001083188.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_001083188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_001199310.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_001199310.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_010715.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
NM_010715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001278.mm.1 |
TC0700000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_001083188.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_001083188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_001199310.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_001199310.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_010715.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
NM_010715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001277.mm.1 |
TC0700000161.mm.1 |
MTA TC |