Assay Details
| Gene Symbol: Zeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011546.3 | NP_035676.1 | 6 - 7 | 2584 | 65 | |
| GenBank mRNA | BC139768.1 | - | 6 - 7 | 2553 | 40111224 | 65 |
| BC139769.1 | - | 6 - 7 | 2553 | 40111227 | 65 | |
| D76432.1 | - | 6 - 7 | 2553 | 65 | ||
| U26259.1 | - | 6 - 7 | 2570 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zeb1
Genomic Map