Assay Details
| Gene Symbol: Tff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011575.2 | NP_035705.1 | 2 - 3 | 264 | 61 | |
| GenBank mRNA | AK008538.1 | - | 2 - 3 | 264 | 61 | |
| BC011042.1 | - | 2 - 3 | 259 | 4216708 | 61 | |
| D38410.1 | - | 2 - 3 | 264 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | D38410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | D38410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388459 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | D38410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | D38410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138044 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08757798 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154853 | - | - |
| Whole Mouse Genome G4122A | A_51_P456208 | D38410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456208 | D38410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456208 | NM_011575.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456208 | AK008538.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102627 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13244430 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13244423 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13244431 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13128951 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13244428 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13244419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13128947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13483750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13186302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13497074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13059782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13128952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13128951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13128946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13341047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13205630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13244424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13341041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13590061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13244434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449659 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417370_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449657 | - | - |
| Mouse Genome 430 2.0 Array | 1417370_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417370_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21786_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21786_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa15620_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700016655.mm.1 | NM_011575.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016655.mm.1 | NM_011575.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016655.mm.1 | TC1700001811.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417370_at | - | - |
| Mouse Expression Set 430 | 1417370_PM_at | - | - |
| Murine Genome U74v2 Set | 100106_PM_at | - | - |
| Murine Genome U74v2 Set | 100106_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417370_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417370_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417370_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417370_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 856010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1580129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2645973 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2645973 | - | - |
| mouse-6 v1.1 (Illumina) | scl50085.4.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50085.4.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2645973 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2645973 | - | - |

Tff3
Genomic Map