| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500006386.mm.1 |
NM_011626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006386.mm.1 |
NM_011626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006386.mm.1 |
TC0500000719.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006394.mm.1 |
NM_011626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006394.mm.1 |
NM_011626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006394.mm.1 |
TC0500000719.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006393.mm.1 |
NM_011626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006393.mm.1 |
NM_011626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006393.mm.1 |
TC0500000719.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006387.mm.1 |
NM_011626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006387.mm.1 |
NM_011626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006387.mm.1 |
TC0500000719.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006388.mm.1 |
NM_011626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006388.mm.1 |
NM_011626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006388.mm.1 |
TC0500000719.mm.1 |
MTA TC |