Assay Details
| Gene Symbol: Tenm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011858.3 | NP_035988.2 | 21 - 22 | 3772 | 68 | |
| GenBank mRNA | AF059485.1 | - | 22 - 23 | 4183 | 68 | |
| AK147329.1 | - | 21 - 22 | 3773 | 68 | ||
| AK147579.1 | - | 21 - 22 | 4072 | 68 | ||
More Information
Set Membership: |
|

Tenm4
Genomic Map