Assay Details
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011942.1 | NP_036072.1 | 4 - 5 | 287 | 71 | |
| GenBank mRNA | AB009653.1 | - | 4 - 5 | 287 | 71 | |
| AK003689.1 | - | 4 - 5 | 254 | 71 | ||
| AK075590.1 | - | 4 - 5 | 279 | 71 | ||
| AK089112.1 | - | 4 - 5 | 279 | 71 | ||
| AK167478.1 | - | 4 - 5 | 274 | 71 | ||
| BC068120.1 | - | 4 - 5 | 313 | 5714650 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypla2
Genomic Map