| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300004213.mm.1 |
NM_012017.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004213.mm.1 |
NM_012017.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004213.mm.1 |
TC1300000647.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004214.mm.1 |
NM_012017.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004214.mm.1 |
NM_012017.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004214.mm.1 |
TC1300000647.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004212.mm.1 |
NM_012017.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004212.mm.1 |
NM_012017.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004212.mm.1 |
TC1300000647.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004211.mm.1 |
NM_012017.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004211.mm.1 |
NM_012017.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004211.mm.1 |
TC1300000647.mm.1 |
MTA TC |