Assay Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013529.3 | NP_038557.1 | 12 - 13 | 1230 | 70 | |
| GenBank mRNA | AB016780.1 | - | 12 - 13 | 1196 | 70 | |
| AK132821.1 | - | 11 - 12 | 1076 | 70 | ||
| BC031928.1 | - | 12 - 13 | 1198 | 4167189 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfpt2
Genomic Map