Assay Details
| Gene Symbol: Gpr162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013533.3 | NP_038561.1 | 2 - 3 | 1550 | 81 | |
| GenBank mRNA | AK134941.1 | - | 2 - 3 | 1364 | 81 | |
| AK144354.1 | - | 1 - 2 | 1319 | 81 | ||
| AK144543.1 | - | 2 - 3 | 1337 | 81 | ||
| AK171145.1 | - | 3 - 4 | 1734 | 81 | ||
| BC031437.1 | - | 2 - 3 | 1550 | 4527410 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr162
Genomic Map