Assay Details
| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013711.3 | NP_038739.2 | 7 - 8 | 629 | 72 | |
| GenBank mRNA | AB027566.1 | - | 7 - 8 | 629 | 72 | |
| AF136399.1 | - | 7 - 8 | 632 | 72 | ||
| AF171053.1 | - | 7 - 8 | 595 | 72 | ||
| AF412308.1 | - | 7 - 8 | 631 | 72 | ||
| AK075828.1 | - | 7 - 8 | 607 | 72 | ||
| AK186982.1 | - | 153 | 72 | |||
| BC013688.1 | - | 7 - 8 | 609 | 3584154 | 72 | |
| BC029344.1 | - | 7 - 8 | 609 | 72 | ||
| BC052157.1 | - | 7 - 8 | 608 | 6506286 | 72 | |
| BC052758.1 | - | 7 - 8 | 608 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd2
Genomic Map