Assay Details
| Gene Symbol: Tcl1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013773.2 | NP_038801.1 | 1 - 2 | 195 | 132 | |
| GenBank mRNA | AF195488.1 | - | 1 - 2 | 195 | 132 | |
| AK136181.1 | - | 1 - 2 | 211 | 132 | ||
| BC052337.1 | - | 1 - 2 | 231 | 30053367 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015275 | BC052337.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015275 | AK136181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015275 | NM_013773.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015275 | AF195488.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82999 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015275 | BC052337.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015275 | AK136181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015275 | NM_013773.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015275 | AF195488.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707627 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736478 | - | - |
| Whole Mouse Genome G4122A | A_52_P289081 | BC052337.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P289081 | AK136181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P289081 | NM_013773.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P289081 | AF195488.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07736470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10029803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707607 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398099 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448981_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398095 | - | - |
| Mouse Genome 430 2.0 Array | 1448981_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448981_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27379_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27379_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007536.mm.1 | NM_013773.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007536.mm.1 | NM_013773.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007536.mm.1 | TC1200001055.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007533.mm.1 | NM_013773.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007533.mm.1 | NM_013773.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007533.mm.1 | TC1200001055.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007534.mm.1 | NM_013773.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007534.mm.1 | NM_013773.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007534.mm.1 | TC1200001055.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007535.mm.1 | NM_013773.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007535.mm.1 | NM_013773.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007535.mm.1 | TC1200001055.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448981_x_at | - | - |
| Mouse Expression Set 430 | 1448981_PM_x_at | - | - |
| Murine Genome U74v2 Set | 96694_PM_at | - | - |
| Murine Genome U74v2 Set | 96694_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448981_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448981_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448981_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448981_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 745465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2633394 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2633394 | - | - |
| mouse-6 v1.1 (Illumina) | scl0027379.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0027379.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2633394 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2633394 | - | - |

Tcl1b1
Genomic Map