Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 5 - 6 | 730 | 139 | |
| GenBank mRNA | AB012725.1 | - | 5 - 6 | 696 | 139 | |
| AK017693.1 | - | 5 - 6 | 731 | 139 | ||
| AK146438.1 | - | 5 - 6 | 711 | 139 | ||
| BC005567.1 | - | 5 - 6 | 721 | 139 | ||
| BC039041.1 | - | 4 - 5 | 2735 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp326
Genomic Map