Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 6 - 7 | 922 | 89 | |
| NR_030777.1 | 4 - 5 | 458 | 89 | |||
| GenBank mRNA | AB012725.1 | - | 6 - 7 | 888 | 89 | |
| AK017693.1 | - | 6 - 7 | 923 | 89 | ||
| BC037055.1 | - | 4 - 5 | 463 | 4948371 | 89 | |
| BC039041.1 | - | 5 - 6 | 2927 | 89 | ||
| BU503985.1 | - | 3 - 4 | 306 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp326
Genomic Map