Assay Details
| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019457.2 | NP_062330.1 | 6 - 7 | 966 | 117 | |
| GenBank mRNA | AF092208.1 | - | 6 - 7 | 911 | 117 | |
| AK077124.1 | - | 6 - 7 | 950 | 117 | ||
| BC046277.1 | - | 6 - 7 | 917 | 6310345 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc6
Genomic Map