Assay Details
| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019457.2 | NP_062330.1 | 9 - 10 | 1191 | 72 | |
| GenBank mRNA | AF092208.1 | - | 9 - 10 | 1136 | 72 | |
| AK077124.1 | - | 9 - 10 | 1175 | 72 | ||
| BC046277.1 | - | 9 - 10 | 1142 | 6310345 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc6
Genomic Map