Assay Details
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019760.3 | NP_062734.1 | 5 - 6 | 722 | 108 | |
| GenBank mRNA | AB029500.1 | - | 5 - 6 | 666 | 108 | |
| AF181685.1 | - | 5 - 6 | 642 | 108 | ||
| AK002847.1 | - | 5 - 6 | 671 | 108 | ||
| AK005203.1 | - | 5 - 6 | 666 | 108 | ||
| AK088340.1 | - | 5 - 6 | 664 | 108 | ||
| AK133668.1 | - | 5 - 6 | 663 | 108 | ||
| AK133975.1 | - | 5 - 6 | 663 | 108 | ||
| AK135359.1 | - | 5 - 6 | 640 | 108 | ||
| AK141574.1 | - | 5 - 6 | 671 | 108 | ||
| AK148745.1 | - | 5 - 6 | 643 | 108 | ||
| AK159280.1 | - | 5 - 6 | 637 | 108 | ||
| AK159436.1 | - | 5 - 6 | 689 | 108 | ||
| AK167817.1 | - | 5 - 6 | 670 | 108 | ||
| BC006940.1 | - | 5 - 6 | 695 | 108 | ||
| BC017148.1 | - | 5 - 6 | 612 | 4506673 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serinc1
Genomic Map