Assay Details
| Gene Symbol: Papolb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019943.2 | NP_064327.1 | 1 | 156 | 97 | |
| GenBank mRNA | AB030729.1 | - | 1 | 156 | 97 | |
| AF039957.1 | - | 1 | 105 | 97 | ||
| BC129896.1 | - | 1 | 134 | 40111521 | 97 | |
| BC129897.1 | - | 1 | 134 | 40111523 | 97 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB030729.1,AF039957.1,BC129896.1,BC129897.1,NM_019943.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152139 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116940 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | AF039957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | BC129896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | BC129897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | NM_019943.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310333 | AB030729.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24152140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24152139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04407091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04985717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04388603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04379269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04848046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24152123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04461530 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535330 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449208_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456915_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535329 | - | - |
| Mouse Genome 430 2.0 Array | 1449208_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456915_at | - | - |
| Mouse Genome 430 2.0 Array | 1456915_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449208_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56522_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56522_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032355.mm.1 | NM_019943.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032355.mm.1 | NM_019943.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032355.mm.1 | TC0500003531.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032356.mm.1 | NM_019943.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032356.mm.1 | NM_019943.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032356.mm.1 | TC0500003531.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449208_PM_at | - | - |
| Mouse Expression Set 430 | 1449208_at | - | - |
| Mouse Expression Set 430 | 1456915_at | - | - |
| Mouse Expression Set 430 | 1456915_PM_at | - | - |
| Murine Genome U74v2 Set | 161033_PM_at | - | - |
| Murine Genome U74v2 Set | 161033_at | - | - |
| Murine Genome U74v2 Set | 169166_r_at | - | - |
| Murine Genome U74v2 Set | 169166_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449208_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449208_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449208_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449208_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456915_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456915_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 509817 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3840450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221282 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221282 | - | - |
| mouse-6 v1.1 (Illumina) | scl25826.1.1_284-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25826.1.1_284-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221282 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221282 | - | - |

Papolb
Genomic Map