Assay Details
| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136063.2 | NP_001129535.1 | 5 - 6 | 737 | 61 | |
| NM_020014.2 | NP_064398.1 | 5 - 6 | 764 | 61 | ||
| GenBank mRNA | AJ276516.1 | - | 5 - 6 | 764 | 61 | |
| AJ276871.1 | - | 5 - 6 | 737 | 61 | ||
| AK090192.1 | - | 3 - 4 | 402 | 61 | ||
| BC111808.1 | - | 5 - 6 | 763 | 61 | ||
| BC141873.1 | - | 5 - 6 | 759 | 61 | ||
| BC141874.1 | - | 5 - 6 | 764 | 40040202 | 61 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
cytokine-mediated signaling pathway
negative regulation of ossification
glial cell-derived neurotrophic factor receptor signaling pathway
receptor activity
coreceptor activity
glial cell-derived neurotrophic factor receptor activity
transmembrane receptor protein tyrosine kinase signaling pathway
cytokine-mediated signaling pathway
negative regulation of ossification
glial cell-derived neurotrophic factor receptor signaling pathway
receptor activity
coreceptor activity
glial cell-derived neurotrophic factor receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632904 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632910 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632909 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632905 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039196 | AJ276871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039196 | AJ276872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039196 | AJ276870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039196 | BC111808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039196 | BC141873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984118 | AJ276871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984118 | AK090192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984118 | BC111808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984118 | BC141873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984118 | BC141874.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984119 | AJ276871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984119 | AK090192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984119 | AJ276870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984119 | BC141873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984119 | BC141874.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63198 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114170 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984119 | AJ276871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984119 | AK090192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984119 | AJ276870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984119 | BC141873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984119 | BC141874.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984118 | AJ276871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984118 | AK090192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984118 | BC111808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984118 | BC141873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984118 | BC141874.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039196 | AJ276871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039196 | AJ276872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039196 | AJ276870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039196 | BC111808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039196 | BC141873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P409839 | AJ276871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P409839 | AK090192.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P409839 | AJ276870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P409839 | BC141873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P409839 | BC141874.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P171197 | AK090192.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P171197 | AB041809.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241426 | AJ276871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241426 | AK090192.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241426 | BC111808.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241426 | BC141873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241426 | BC141874.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21632922 | - | - |
| Whole Mouse Genome G4122A | A_52_P409839 | AJ276871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409839 | AK090192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409839 | AJ276870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409839 | BC141873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409839 | BC141874.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241426 | AJ276871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241426 | AK090192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241426 | BC111808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241426 | BC141873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241426 | BC141874.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P171197 | AK090192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P171197 | AB041809.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108760 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01251035 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01251042 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01251024 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01844051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02248136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02248139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632904 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632910 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21632909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21632920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01895587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02048508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01251024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01844051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01251027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01895597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01229130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01895598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01251031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02002190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21632933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01251036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10487796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487788 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450835_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10487787 | - | - |
| Mouse Genome 430 2.0 Array | 1450835_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450835_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14588_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14588_PM_at | - | - |
| Mouse Expression Set 430 | 1450835_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450835_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450835_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450835_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450835_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450835_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4590746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738522 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2738522 | - | - |
| mouse-6 v1.1 (Illumina) | scl014588.3_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590746 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014588.3_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2738522 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738522 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014588.3_20-S | - | - |

Gfra4
Genomic Map