| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200046215.mm.1 |
NM_001141933.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046215.mm.1 |
NM_001141933.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046215.mm.1 |
NM_021426.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046215.mm.1 |
NM_021426.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046215.mm.1 |
TC0200005426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200046210.mm.1 |
NM_001141933.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046210.mm.1 |
NM_001141933.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046210.mm.1 |
NM_021426.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046210.mm.1 |
NM_021426.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046210.mm.1 |
TC0200005426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200046214.mm.1 |
NM_001141933.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046214.mm.1 |
NM_001141933.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046214.mm.1 |
NM_021426.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046214.mm.1 |
NM_021426.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046214.mm.1 |
TC0200005426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200046213.mm.1 |
NM_001141933.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046213.mm.1 |
NM_001141933.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046213.mm.1 |
NM_021426.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046213.mm.1 |
NM_021426.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200046213.mm.1 |
TC0200005426.mm.1 |
MTA TC |